Anti-STX18 Rabbit Polyclonal Antibody

Supplier: Proteintech
16013-1-AP-150UL
10095-526EA 402 USD
10095-526
Anti-STX18 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Liver Tissue, 1:500-1:5000; IHC: Human endometrial cancer Tissue, 1:20-1:200

Type: Primary
Antigen: STX18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR