Anti-NBR1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16004-1-AP-150UL
10090-844EA 537.7 USD
10090-844
Anti-NBR1 Rabbit Polyclonal Antibody
Antibodies
NBR1, also named as 1A13B, KIAA0049 and M17S2, acts probably as a receptor for selective autophagosomal degradation of ubiquitinated targets. NBR1 and P62 can bind to autophagic effector proteins (Atg8 in yeast, MAP1LC3 protein family in mammals) anchored in the membrane of autophagosomes. It is a highly conserved multidomain scaffold protein with proposed roles in endocytic trafficking and selective autophagy. NBR1 is a novel PB1 adapter in Th2 differentiation and asthma. It functions as an autophagy receptor involved in targeting ubiquitinated proteins for degradation. It also has a dual role as a scaffold protein to regulate growth-factor receptor and downstream signaling pathways.

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human breast cancer Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100; IP: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: NBR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR