Anti-BAAT Rabbit Polyclonal Antibody

Supplier: Proteintech
15990-1-AP-150UL
10083-472EA 402 USD
10083-472
Anti-BAAT Rabbit Polyclonal Antibody
Antibodies
BAAT is bile acid-CoA:amino acid N-acyltransferase which involved in in bile acid metabolism. BAAT acts as an acyl-CoA thioesterase that regulates intracellular levels of free fatty acids. BAAT protein showes 46-50 kDa in WB detection.

Western Blot: Human Liver Tissue, 1:200-1:2000; IF: HepG2 Cells, 1:10-1:100; IHC: Human hepatocirrhosis Tissue, 1:20-1:200

Type: Primary
Antigen: BAAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR