Anti-ACOT7 Rabbit Polyclonal Antibody

Supplier: Proteintech
15972-1-AP-150UL
10082-438EA 459.7 USD
10082-438
Anti-ACOT7 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Brain Tissue, 1:1000-1:10000; IHC: Human Brain, 1:20-1:200

Type: Primary
Antigen: ACOT7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR