Anti-CLIC3 Rabbit Polyclonal Antibody

Supplier: Proteintech
15971-1-AP-150UL
10084-862EA 402 USD
10084-862
Anti-CLIC3 Rabbit Polyclonal Antibody
Antibodies
Chloride channels are a diverse group of proteins that regulate fundamental cellular processes including stabilization of cell membrane potential, transepithelial transport, maintenance of intracellular pH, and regulation of cell volume . CLIC3 (chloride intracellular channel protein 3) belongs to the chloride channel CLIC family which has seven members based on sequence homology to the p64 chloride channels of bovine kidney microsomes. CLIC proteins lack the membrane-spanning domains characteristic of channel proteins and are largely intracellular. CLIC3 is predominantly localized in the nucleus and has a high expression level in placenta . It associates with the C-terminal of ERK7 and may participate in cellular growth control.

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human Placenta Tissue, 1:20-1:200; IF: MCF-7 Cells, 1:10-1:100

Type: Primary
Antigen: CLIC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR