Anti-CSTA Rabbit Polyclonal Antibody

Supplier: Proteintech
15962-1-AP-150UL
10084-642EA 402 USD
10084-642
Anti-CSTA Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Liver Tissue, 1:200-1:2000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: CSTA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR