Anti-MBD5 Rabbit Polyclonal Antibody

Supplier: Proteintech
15961-1-AP-150UL
10090-010EA 402 USD
10090-010
Anti-MBD5 Rabbit Polyclonal Antibody
Antibodies
In vertebrates, cytosine methylation in DNA is one of the major epigenetic modifications, which regulates many cellular events, including developmental gene expression, X chromosome inactivation, genome defense, and genomic imprinting . DNA methylation exerts regulatory functions by recruiting specific binding proteins that contain a highly conserved methyl-CpG binding domain (MBD) . Methyl-CpG binding domain protein 5 (MBD5) belongs to the MBD family proteins, which play central roles in transcriptional regulation and development. It is a candidate gene involved in human 2q23.1 microdeletion syndrome .

Western Blot: Mouse Heart Tissue, 1:200-1:2000

Type: Primary
Antigen: MBD5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR