Anti-PIEZO1 Rabbit Polyclonal Antibody

Supplier: Proteintech
15939-1-AP-150UL
10086-810EA 537.7 USD
10086-810
Anti-PIEZO1 Rabbit Polyclonal Antibody
Antibodies
Mechanotransduction, the conversion of mechanical force into biological signals, is a fundamental physiologic process of mammalian cells which influences many critical processes including embryonic development, tactile, pain, and auditory sensation, regulation of vascular tone, flow sensing in the kidney, and muscle and tendon stretch. FAM38A, also known as PIEZO1, has recently been identified as a mechanotransduction protein that gets involved in mechanosensation and stretch-activated cation channel activation. Fam38A also plays a key role in epithelial cell adhesion by maintaining integrin activation through R-Ras recruitment to the ER. Mutations in gene encoding PIEZO1 are associated with hereditary xerocytosis. Piezo1 also regulates extrusion to maintain homeostatic epithelial cell numbers.

Western Blot: HepG2 Cells, 1:500-1:5000

Type: Primary
Antigen: PIEZO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR