Anti-CTP Synthase Rabbit Polyclonal Antibody

Supplier: Proteintech
15914-1-AP-150UL
10085-322EA 402 USD
10085-322
Anti-CTP Synthase Rabbit Polyclonal Antibody
Antibodies
CTP synthase(CTPS) is also named as CTPS1 and belongs to the CTP synthase family. It catalyses the ATP-dependent formation of CTP from UTP using either L-glutamine or NH3 as the nitrogen source. It is important in the biosynthesis of phospholipids and nucleic acids, and plays a key role in cell growth, development, and tumorigenesis.

Western Blot: HeLa Cells, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100; FC: HepG2 Cells, N/A

Type: Primary
Antigen: CTP Synthase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR