Anti-RAB11 Rabbit Polyclonal Antibody

Supplier: Proteintech
15903-1-AP-150UL
10092-936EA 453 USD
10092-936
Anti-RAB11 Rabbit Polyclonal Antibody
Antibodies
RAB11 has two isoform genes RAB11A(YL8) and RAB11B(YPT3), and belongs to the small GTPase superfamily and Rab family. It regulates endocytic recycling. RAB11 may exert its functions by interacting with multiple effector proteins in different complexes. It acts as a major regulator of membrane delivery during cytokinesis. RAB11 regulates the recycling of FCGRT (receptor of Fc region of monomeric Ig G) to basolateral membranes. This antibody can recognize both RAB11A(24kd) and RAB11B(24kd).

Western Blot: Mouse spleen Tissue, 1:500-1:5000

Type: Primary
Antigen: RAB11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR