Anti-Creatine Kinase Rabbit Polyclonal Antibody

Supplier: Proteintech
15891-1-AP-150UL
10084-804EA 402 USD
10084-804
Anti-Creatine Kinase Rabbit Polyclonal Antibody
Antibodies
Creatine Kinase, an enzyme important for energy metabolism in cells of high and fluctuating energy requirements, catalyses the reversible transfer of a phosphoryl goup from phosphocreatine to ADP. CK isoenzymes(muscle, brain, sarcomeric, Ubiquitous-type) play a central role in energy transduction in tissues with large, fluctuating energy demands, such as skeletal muscle, heart, brain and spermatozoa. Inactivation of creatine kinase by gliotoxin was accompanied by the formation of a 37-kDa form of the enzyme.This oxidized form of creatine kinase was rapidly reconverted to the 42-kDa species by the addition of reducing agents concomitant with restoration of activity..

Western Blot: Mouse Skeletal Muscle Tissue, 1:1000-1:10000

Type: Primary
Antigen: Creatine Kinase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR