Anti-PPP2R1A Rabbit Polyclonal Antibody

Supplier: Proteintech
15882-1-AP-150UL
10092-706EA 402 USD
10092-706
Anti-PPP2R1A Rabbit Polyclonal Antibody
Antibodies
PPP2R1A belongs to the phosphatase 2A regulatory subunit A family. PPP2R1A encodes the α-isoform of the scaffolding subunit of the serine/threonine protein phosphatase 2A (PP2A) holoenzyme. This tumour suppressor complex is involved in growth and survival pathways and PPP2R1A mutations in endometrial high-grade serous carcinomas are a frequent and potentially targetable feature of this disease. The PPP2R1A gene has 86% identity with the PPP2R1B gene.

Western Blot: HepG2 Cells, 1:200-1:2000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: PPP2R1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR