Anti-SPANXE Rabbit Polyclonal Antibody

Supplier: Proteintech
15870-1-AP-150UL
10095-060EA 402 USD
10095-060
Anti-SPANXE Rabbit Polyclonal Antibody
Antibodies
SPANXE (Sperm protein associated with the nucleus on the X chromosome E), also known as SPANX A2 or SPANX C, is a member of the SPANX family. The SPANX genes encode differentially expressed testis-specific proteins that localize to various subcellular compartments. This particular gene encodes a sperm protein that contains a consensus nuclear localization signal but, although a role in spermatogenesis is suggested, the specific function of this family member has not yet been determined.

Western Blot:A375 Cells, 1:200-1:1000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: SPANXE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR