Anti-PSMC6 Rabbit Polyclonal Antibody

Supplier: Proteintech
15839-1-AP-150UL
10092-922EA 402 USD
10092-922
Anti-PSMC6 Rabbit Polyclonal Antibody
Antibodies
PSMC6(proteasome (prosome, macropain) 26S subunit, ATPase, 6) is also named as CADP44, MGC12520, P44, SUG2, p42 and belongs to the AAA ATPase family. It is a component of the 26S protease, which is involved in the ATP-dependent degradation of ubiquitinated proteins. The regulatory (or ATPase) complex confers ATP dependency and substrate specificity to the 26S complex. The gene encodes a 44 kDa PSMC6.

Western Blot: Human Brain Tissue, 1:1000-1:10000

Type: Primary
Antigen: PSMC6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR