Anti-GSTT1 Rabbit Polyclonal Antibody

Supplier: Proteintech
15838-1-AP-150UL
10087-942EA 402 USD
10087-942
Anti-GSTT1 Rabbit Polyclonal Antibody
Antibodies
GSTT1 belongs to the GST superfamily and Theta family. It is a conjugation of reduced glutathione to a wide number of exogenous and endogenous hydrophobic electrophiles. GSTT1 acts on 1,2-epoxy-3-(4-nitrophenoxy)propane, phenethylisothiocyanate 4-nitrobenzyl chloride and 4-nitrophenethyl bromide. It displays glutathione peroxidase activity with cumene hydroperoxide. GSTM1 and GSTT1 has been suggested as a risk factor in various cancers. GSTT1 is important in the detoxification of unidentified xenobiotics in the large intestine.

Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: GSTT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR