Anti-Carboxypeptidase A1 Rabbit Polyclonal Antibody

Supplier: Proteintech
15836-1-AP-150UL
10085-090EA 402 USD
10085-090
Anti-Carboxypeptidase A1 Rabbit Polyclonal Antibody
Antibodies
CPA1(Carboxypeptidase A1) is also named as CPA and belongs to the the peptidase M14 family. CPA1 is one of the genes whose expression in the pancreas and the protein which it encodes is synthesized as an inactive precursor, proCPA1, which is processed to the active enzyme by the proteolytic removal of the 95-amino acid N-terminal prodomain. The mature CPA1 is not secreted and is trapped and degraded intracellularly. This antibody is speicific to CPA1.

Western Blot: Mouse pancreas Tissue, 1:500-1:5000; IHC: Human Placenta Tissue, 1:20-1:200

Type: Primary
Antigen: Carboxypeptidase A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR