Anti-domain-I-of-FIZ-1 Rabbit Polyclonal Antibody

Supplier: Proteintech
15826-1-AP-150UL
10081-774EA 402 USD
10081-774
Anti-domain-I-of-FIZ-1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Kidney Tissue, 1:200-1:2000; IF: HepG2 Cells, 1:10-1:100; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: domain-I-of-FIZ-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR