Anti-HMMR Rabbit Polyclonal Antibody

Supplier: Proteintech
15819-1-AP-150UL
10093-636EA 402 USD
10093-636
Anti-HMMR Rabbit Polyclonal Antibody
Antibodies
HMMR, also named as RHAMM, is a hyaluronan receptor expressed in various tissues. It is involved in regulation of focal contacts and subsequent motility, thus to play function in wound healing and tumor metastasis. The protein was expressed as a 70kd one in many tissues. This antibody is specific for rat HAMM, not react with Human and Mouse HAMM.

Western Blot:NIH/3T3 Cells, 1:200-1:2000

Type: Primary
Antigen: HMMR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR