Anti-PRDX1 Rabbit Polyclonal Antibody

Supplier: Proteintech
15816-1-AP-150UL
10081-408EA 537.7 USD
10081-408
Anti-PRDX1 Rabbit Polyclonal Antibody
Antibodies
PRDX1(Peroxiredoxin-1) is a thiol reductase that plays critical roles in oxidative and thermal stress defense mechanisms through its abilities to metabolize H2O2 and act as a molecular chaperone, respectively.PRDX1 also reduces intramolecular cystine bridges .It can form a dimer.It also can be phosphorylated on Thr-90 during the M-phase, which leads to a more than 80% decrease in enzymatic activity.

Western Blot: HEK-293 Cells, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; IP: HeLa Cells, 1:500-1:5000; FC: HepG2 Cells, N/A

Type: Primary
Antigen: PRDX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR