Anti-UBE2O Rabbit Polyclonal Antibody

Supplier: Proteintech
15812-1-AP-150UL
10085-608EA 402 USD
10085-608
Anti-UBE2O Rabbit Polyclonal Antibody
Antibodies
UBE2O, also named as KIAA1734, belongs to the ubiquitin-conjugating enzyme family. It catalyzes the covalent attachment of ubiquitin to other proteins.

Western Blot: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: UBE2O
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR