Anti-NHP2L1 Rabbit Polyclonal Antibody

Supplier: Proteintech
15802-1-AP-150UL
10091-098EA 537.7 USD
10091-098
Anti-NHP2L1 Rabbit Polyclonal Antibody
Antibodies
NHP2L1, belongs to the ribosomal protein L7Ae family, is a component of the spliceosome complex. It is recruited to introns following the attachment of U4 and U6 RNAs, and is required for subsequent recruitment of PRP31 and activation of the spliceosomal complex . It binds to the 5'-stem-loop of U4 snRNA and may have a role in the late stage of spliceosome assembly. The protein undergoes a conformational change upon RNA-binding .

Western Blot: HeLa Cells, 1:500-1:5000; IF: HeLa Cells, 1:10-1:100; IHC: Human cervical cancer Tissue, 1:20-1:200

Type: Primary
Antigen: NHP2L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR