Anti-CIP29 Rabbit Polyclonal Antibody

Supplier: Proteintech
15798-1-AP-150UL
10084-592EA 402 USD
10084-592
Anti-CIP29 Rabbit Polyclonal Antibody
Antibodies
Western Blot: MCF7 Cells, 1:500-1:5000

Type: Primary
Antigen: CIP29
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR