Anti-RBM41 Rabbit Polyclonal Antibody

Supplier: Proteintech
15784-1-AP-150UL
10093-420EA 402 USD
10093-420
Anti-RBM41 Rabbit Polyclonal Antibody
Antibodies
Western Blot:SGC-7901 Cells, 1:500-1:5000; IHC: Human endometrial cancer, 1:20-1:200; IF: HepG2, 1:20-1:200

Type: Primary
Antigen: RBM41
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR