Anti-ALDH3B2 Rabbit Polyclonal Antibody

Supplier: Proteintech
15746-1-AP-150UL
10082-764EA 402 USD
10082-764
Anti-ALDH3B2 Rabbit Polyclonal Antibody
Antibodies
ALDH3B2(aldehyde dehydrogenase family 3 member B2) is also named as ALDH8(aldehyde dehydrogenase 8) and belongs to the aldehyde dehydrogenase family. The ALDH3B2 gene contains an in‐frame stop codon at the seventeenth codon position from the first methionine,suggesting that ALDH3B2 may be a pseudogene and encodes the human salivary gland and placental ALDH. This protein shares 83% sequence identity with ALDH3B1.

Western Blot: Mouse lung Tissue, 1:500-1:5000; IHC: Human adrenal gland, 1:20-1:200; IF: Hela, 1:10-1:100

Type: Primary
Antigen: ALDH3B2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR