Anti-TRIM14 Rabbit Polyclonal Antibody

Supplier: Proteintech
15742-1-AP-150UL
10096-314EA 402 USD
10096-314
Anti-TRIM14 Rabbit Polyclonal Antibody
Antibodies
Tripartite motif-containing 14(TRIM14) belongs to the TRIM protein family that contains 3 zinc-binding domains, a RING, a B-box type1 and a B-box type2 and plays key roles in cellular proliferation, differentiation, development, transformation, and immune response mechanisms . TRIM4 inhibits the transcriptional activity of SPI1 in a dose-dependent manner. It may participate in the early stages of differentiation of embryonic stem cells, leading to the formation of embryoid bodies.

Western Blot:L02 Cells, 1:500-1:5000; IHC: Human Liver, 1:20-1:200; IF: Hela, 1:10-1:100

Type: Primary
Antigen: TRIM14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR