Anti-HLA-DRB4 Rabbit Polyclonal Antibody

Supplier: Proteintech
15733-1-AP-150UL
10088-306EA 402 USD
10088-306
Anti-HLA-DRB4 Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:200-1:2000

Type: Primary
Antigen: HLA-DRB4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR