Anti-SNX9 Rabbit Polyclonal Antibody

Supplier: Proteintech
15721-1-AP-150UL
10094-982EA 402 USD
10094-982
Anti-SNX9 Rabbit Polyclonal Antibody
Antibodies
Sorting nexins are a diverse group of cytoplasmic and membrane-associated proteins that are classified by the presence of a phospholipid-binding motif-the PX domain . They are involved in endocytosis and protein trafficking. SNX9 (Sorting nexin-9, also known as SH3PX1) was originally identified as a protein that interacted with the metalloproteinases ADAM9 and ADAM15. It contains a PX and an Sec homology 3 (SH3) domain. SNX9 functions in clathrin-mediated endocytosis and clathrin-independent, actin-dependent fluid-phase endocytosis . On SDS-PAGE, SNX9 migrates at a molecular weight (70-78 kDa) higher than its calculated molecular mass of 67 kDa .

Western Blot: HeLa Cells, 1:1000-1:10000; IF: HeLa Cells, 1:20-1:200; IP: Mouse Heart Tissue, 1:500-1:5000

Type: Primary
Antigen: SNX9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR