Anti-RIMBP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
15716-1-AP-150UL
10093-678EA 402 USD
10093-678
Anti-RIMBP2 Rabbit Polyclonal Antibody
Antibodies
The presynaptic active zone mediates synaptic vesicle exocytosis at a synapse. RIM proteins are active zone scaffolding molecules that--among others--mediate vesicle priming and directly or indirectly interact with most other essential presynaptic proteins. RIMBP2 encodes RIMS-binding protein 2 belonging to RIMBP family. RIMBP2 is supposed to be localized at cell membrane especially synaptic plasma membrane. By interacting with RIMS1, RIMS2, CACNA1D and CACNA1B via the SH3 domains, RIMBP2 mediates synaptic transmission as bifunctional linker.

Western Blot: Mouse Kidney Tissue, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: RIMBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR