Anti-AMPD2 Rabbit Polyclonal Antibody

Supplier: Proteintech
15710-1-AP-150UL
10082-652EA 402 USD
10082-652
Anti-AMPD2 Rabbit Polyclonal Antibody
Antibodies
Adenosine monophosphate deaminase (AMPD) catalyzes the deamination of AMP to IMP and plays an important role in the purine nucleotide cycle. Three AMPDs are present in mammals. AMPD2 is the predominant form in smooth muscle, non-muscle tissue, embryonic muscle and undifferentiated myoblasts. At least four transcripts of AMPD2 were reported. The longest one encodes a protein with calculated MW of 101kda.

Western Blot: Human Liver Tissue, 1:500-1:5000

Type: Primary
Antigen: AMPD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR