Anti-ACSL5 Rabbit Polyclonal Antibody

Supplier: Proteintech
15708-1-AP-150UL
10082-462EA 402 USD
10082-462
Anti-ACSL5 Rabbit Polyclonal Antibody
Antibodies
Acyl-CoA synthetase 5, a member of the ACSL gene family that catalyzes the activation of long-chain fatty acids for lipid biosynthesis, is the only ACSL isoform that is both, located on mitochondria and functionally involved in enterocyte apoptosis.ACSL5 could play a role in promoting fatty acid-induced lipoapoptosis in hepatocytes as important mechanism in fatty liver-related disorders. ACSL5 is most abundant in liver, brown adipose tissue, and intestine and is located on both the mitochondrial membrane and endoplasmic reticulum. It has 3 isoforms produced by alternative splicing.

Western Blot: Mouse Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: ACSL5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR