Anti-L2HGDH Rabbit Polyclonal Antibody

Supplier: Proteintech
15707-1-AP-150UL
10089-408EA 402 USD
10089-408
Anti-L2HGDH Rabbit Polyclonal Antibody
Antibodies
L2HGDH(L-2-hydroxyglutarate dehydrogenase, mitochondrial) is also named as duranin, C14orf160 and belongs to the L2HGDH family.T he putative L2HGDH is predicted to be targeted to the mitochondria where its mitochondrial targeting sequence is presumably removed. Defects in L2HGDH are the cause of L-2-hydroxyglutaric aciduria (L2HGA). It has 2 isoforms produced by alternative splicing with the molecular weight of 50 kDa and 48 kDa.

Western Blot: Mouse small intestine Tissue, 1:200-1:2000; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: L2HGDH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR