Anti-IGFBP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
15699-1-AP-150UL
10088-662EA 402 USD
10088-662
Anti-IGFBP2 Rabbit Polyclonal Antibody
Antibodies
Insulin-like growth factors (IGFs) and their binding proteins (IGFBPs) have important metabolic roles in the human body. IGFBP-2 is a 34 kD IGFBP protein with degarded fragments of 24 kD, 19kd and 15kd. The levels of IGFBP2 are elevated during progression of many human cancers. IGFBP2 is expressed in invasive PAc, whereas its expression in HG-PIN is low. It can be as an Immunohistochemical Marker for Prostatic Adenocarcinoma.

Recommended dilution: WB : 1:200-1:2000; IHC : 1:20-1:200; IF : 1:10-1:100

Type: Primary
Antigen: IGFBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR