Anti-TK1 Rabbit Polyclonal Antibody

Supplier: Proteintech
15691-1-AP-150UL
10096-036EA 402 USD
10096-036
Anti-TK1 Rabbit Polyclonal Antibody
Antibodies
TK1(Thymidine kinase, cytosolic) belongs to the thymidine kinase family and is a regulatory enzyme that can interchange between its dimeric and tetrameric forms, which have low and high catalytic efficiencies, respectively. TK1 is involved in DNA synthesis and an important proliferation marker.

Western Blot: HEK-293 Cells, 1:200-1:2000; IF: Hela Cells, 1:10-1:100; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: TK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR