Anti-AP2B1 Rabbit Polyclonal Antibody

Supplier: Proteintech
15690-1-AP-150UL
10082-966EA 402 USD
10082-966
Anti-AP2B1 Rabbit Polyclonal Antibody
Antibodies
AP2B1 is a component of the adaptor protein complex 2 (AP-2). AP complexes are cytosolic heterotetramers that mediate the sorting of membrane proteins in the secretory and endocytic pathways. AP complexes are involved in the formation of clathrin-coated vesicles (CCVs) by recruiting the scaffold protein, clathrin. AP complexes also play a pivotal role in the cargo selection by recognizing the sorting signals within the cytoplasmic tail of integral membrane proteins. AP-2 works on the plasma membrane to internalize cargo in clathrin-mediated endocytosis.

Western Blot:K-562 Cells, 1:1000-1:10000; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: AP2B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR