Anti-DLG5 Rabbit Polyclonal Antibody

Supplier: Proteintech
15687-1-AP-150UL
10085-896EA 402 USD
10085-896
Anti-DLG5 Rabbit Polyclonal Antibody
Antibodies
Western Blot:PC-3 Cells, 1:200-1:2000

Type: Primary
Antigen: DLG5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR