Anti-REEP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
15684-1-AP-150UL
10093-500EA 402 USD
10093-500
Anti-REEP2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Skeletal Muscle Tissue, 1:500-1:5000

Type: Primary
Antigen: REEP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR