Anti-ARHGAP9 Rabbit Polyclonal Antibody

Supplier: Proteintech
15665-1-AP-150UL
10083-084EA 402 USD
10083-084
Anti-ARHGAP9 Rabbit Polyclonal Antibody
Antibodies
ARHGAP9 belongs to RHO family of small GTPases, which inactivate RHO GTPases by stimulating their intrinsic GTPase activity. Several ARHGAP9 transcript isoforms were reported: including calculated mass of 84kda, 70kd and 64kda .

Western Blot: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: ARHGAP9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR