Anti-CPA2 Rabbit Polyclonal Antibody

Supplier: Proteintech
15626-1-AP-150UL
10085-092EA 402 USD
10085-092
Anti-CPA2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse pancreas Tissue, 1:1000-1:10000; IHC: Human pancreas Tissue, 1:20-1:200

Type: Primary
Antigen: CPA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR