Anti-FCGR2A/ CD32a Rabbit Polyclonal Antibody

Supplier: Proteintech
15625-1-AP-150UL
10084-328EA 402 USD
10084-328
Anti-FCGR2A/ CD32a Rabbit Polyclonal Antibody
Antibodies
IgG Fc receptors (FcγRs) play important roles in immune responses. Low affinity IgG Fc receptor Fc gamma RIIa (FCGR2A, CD32a) is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes.

Western Blot:Jurkat Cells, 1:500-1:5000

Type: Primary
Antigen: FCGR2A/ CD32a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR