Anti-GABRD Rabbit Polyclonal Antibody

Supplier: Proteintech
15623-1-AP-150UL
10087-236EA 402 USD
10087-236
Anti-GABRD Rabbit Polyclonal Antibody
Antibodies
Gamma-aminobutyric acid (GABA) is the major inhibitory neurotransmitter in the mammalian brain, The GABRD gene encodes the delta subunit of the ligand-gated chloride channel for gamma-aminobutyric acid (GABA). This protein migrates as a 55-65kd protein in SDS-PAGE.

Western Blot: Human Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: GABRD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR