Anti-NSFL1C Rabbit Polyclonal Antibody

Supplier: Proteintech
15620-1-AP-150UL
10091-320EA 402 USD
10091-320
Anti-NSFL1C Rabbit Polyclonal Antibody
Antibodies
Western Blot:A431 Cells, 1:200-1:2000

Type: Primary
Antigen: NSFL1C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR