Anti-CBR3 Rabbit Polyclonal Antibody

Supplier: Proteintech
15619-1-AP-150UL
10084-134EA 402 USD
10084-134
Anti-CBR3 Rabbit Polyclonal Antibody
Antibodies
CBR3(Carbonyl reductase [NADPH] 3), which Belongs to the short-chain dehydrogenases/reductases (SDR) family, has low NADPH-dependent oxidoreductase activity towards 4-benzoylpyridine and menadione (in vitro). The gene encodes a 31 kDa protein that can catalyze the reduction of many endogenous and xenobiotic carbonyl compounds, including steroids and prostaglandins. The deduced 277-amino acid CBR3 protein is 84% similar to the CBR1 protein.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human Colon cancer Tissue, 1:20-1:200

Type: Primary
Antigen: CBR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR