Anti-SELS Rabbit Polyclonal Antibody

Supplier: Proteintech
15591-1-AP-150UL
10094-286EA 402 USD
10094-286
Anti-SELS Rabbit Polyclonal Antibody
Antibodies
SELS, also named as VIMP and SEPS1, plays an important role in the production of inflammatory cytokines and its expression is activated by endoplasmic reticulum (ER) stress. It probably acts by serving as a linker between DERL1 and VCP.

Western Blot: HeLa Cells, 1:500-1:5000; IP: HepG2 Cells, 1:200-1:2000

Type: Primary
Antigen: SELS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR