Anti-PAIP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
15583-1-AP-150UL
10091-754EA 402 USD
10091-754
Anti-PAIP2 Rabbit Polyclonal Antibody
Antibodies
Western Blot:A431 Cells, 1:200-1:2000; IF: Hela, 1:10-1:100

Type: Primary
Antigen: PAIP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR