Anti-GMCL1 Rabbit Polyclonal Antibody

Supplier: Proteintech
15575-1-AP-150UL
10087-652EA 402 USD
10087-652
Anti-GMCL1 Rabbit Polyclonal Antibody
Antibodies
GMCL1, also named as GCL, functions in spermatogenesis. It enhances the degradation of MDM2 and increases the amount of p53 probably by modulating the nucleocytoplasmic transport. In WB test, the antibody recognizes 55 kDa band in our detection.

Western Blot: Human Heart Tissue, 1:500-1:5000

Type: Primary
Antigen: GMCL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR