Anti-NDUFC2 Rabbit Polyclonal Antibody

Supplier: Proteintech
15573-1-AP-150UL
10090-976EA 402 USD
10090-976
Anti-NDUFC2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Kidney Tissue, 1:500-1:5000; IHC: Human Kidney, 1:20-1:200

Type: Primary
Antigen: NDUFC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR