Anti-DIRAS2 Rabbit Polyclonal Antibody

Supplier: Proteintech
15557-1-AP-150UL
10085-866EA 402 USD
10085-866
Anti-DIRAS2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Brain Tissue, 1:500-1:5000; IF: Hela Cells, 1:10-1:100; FC: HeLa Cells, N/A

Type: Primary
Antigen: DIRAS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR