Anti-PPM1G Rabbit Polyclonal Antibody

Supplier: Proteintech
15532-1-AP-150UL
10092-668EA 402 USD
10092-668
Anti-PPM1G Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse lung Tissue, 1:200-1:2000; IHC: Human Testis, 1:20-1:200

Type: Primary
Antigen: PPM1G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR