Anti-USP44 Rabbit Polyclonal Antibody

Supplier: Proteintech
15521-1-AP-150UL
10096-878EA 402 USD
10096-878
Anti-USP44 Rabbit Polyclonal Antibody
Antibodies
USP44(Ubiquitin carboxyl-terminal hydrolase 44) belongs to a large family of cysteine proteases that function as deubiquitinating enzymes. It has a ZnF-UBP (zinc-finger ubiquitin-specific protease) domain and conserved cysteine, histidine and asparagine/aspartic acid residues characteristic of deubiquitinating enzymes. And USP44 can cleave ubiquitin from ubiquitinated substrates both in vitro and in vivo. The expression of endogenous mouse or human USP44 is very low. A localization study of USP44 showed its predominant expression in the nucleus.

Western Blot: Human Placenta Tissue, 1:500-1:5000

Type: Primary
Antigen: USP44
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR