Anti-MSH2 Rabbit Polyclonal Antibody

Supplier: Proteintech
15520-1-AP-150UL
10089-466EA 402 USD
10089-466
Anti-MSH2 Rabbit Polyclonal Antibody
Antibodies
MSH2, named for its homologous to the E.coli MutS gene, takes part in DNA mismatch repair(MMR) to maintain the stability of DNA. MSH2 contains a helix-turn-helix domain, which responds for binding to DNA. Also, MSH2 has a direct role in mutation avoidance and micro-satellite stability in cell.

Western Blot: HeLa Cells, 1:1000-1:10000; IHC: Human Colon cancer, 1:20-1:200; IF: Hela, 1:10-1:100

Type: Primary
Antigen: MSH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR